Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAPDH 293T Cell Overexpression Lysate (Denatured), Abnova
Cell transient overexpression lysate for GAPDH
Manufacturer: Abnova Corporation H00002597T01
Description
The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD).
- Glyceraldehyde-3-phosphate dehydrogenase
- Transfected cell line: 293T
- Plasmid: pCMV-GAPDH full-length
- Host: Human
- Theoretical molecular weight: 36.1kDa
- Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western Blotting
Specifications
Western Blot | |
GAPDH 293T Cell Transient Overexpression Lysate(Denatured) | |
36.1 | |
pCMV-GAPDH full-length | |
100 μL | |
Human |
293T | |
NM_002046 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-GAPDH antibody (H00002597-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title