Learn More
GAPDH Mouse anti-Human, Clone: B903M, Abnova™
Mouse monoclonal antibody raised against human GAPDH.
Manufacturer:  Abnova Corporation MAB12720
Catalog No. 89-934-186
Add to cart
Description
Description
The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. Many pseudogenes similar to this locus are present in the human genome. [provided by RefSeq]
Specifications
Specifications
GAPDH
B903M
Mouse monoclonal antibody raised against human GAPDH.
In PBS, pH 7.4 (0.09% sodium azide).
G3PD/GAPD/MGC88685
Mouse
IgG1
250 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunocytochemistry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
GAPDH
Human GAPDH
Protein A purification
RUO
Primary
2597