Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH (P. falciparum) Mouse anti-Plasmodium falciparum, Unconjugated, Clone: 1.4, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $380.50

Specifications

Antigen GAPDH (P. falciparum)
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS19461MI
View Documents
MilliporeSigma
MABS1946100UG
100 μg
Each for $380.50
Add to cart
 
MABS19462MI
View Documents
MilliporeSigma
MABS194625UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

GAPDH (P. falciparum) Monoclonal antibody specifically detects GAPDH (P. falciparum) in Plasmodium falciparum samples. It is validated for Immunofluorescence, Western Blotting.
Specifications

Specifications

GAPDH (P. falciparum)
Unconjugated
Mouse
Glyceraldehyde-3-phosphate dehydrogenase;EC 1.2.1.12
His-tagged full length recombinant Glyceraldehyde-3-phosphate dehydrogenase from Plasmodium falciparum.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunofluorescence, Western Blot
Purified
Plasmodium
PF3D7_1462800
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit