Learn More
Tocris Bioscience™ Garcinol
PCAF/p300 inhibitor; anticancer
Manufacturer:  Tocris Bioscience™ 4827/10
Catalog No. 48-271-0
Add to cart
This item is not returnable. View return policy
Description
Description
Inhibitor of PCAF and p300 histone acetyltransferase (HAT) activity (IC50 values are 5 and 7 μM, respectively). Promotes expansion of human HSCs ex vivo. Causes reversal of EMT and inhibits β-catenin in breast cancer cell lines MDA-MB-231 and BT-549.
Specifications
Specifications
Garcinol
>98
10mg
Garcinol
602.8