Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GARS Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI4G4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270794AF405

Catalog No. N270794A405


Add to cart

Description

Description

GARS Monoclonal antibody specifically detects GARS in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

GARS
OTI4G4
Immunohistochemistry
Affinity Purified, Conjugated
GARS
Full length recombinant protein of human GARS (NP_002038) produced in HEK293T cells.
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
2617.0
Immunohistochemistry
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
AP-4-A synthetase, Charcot-Marie-Tooth neuropathy 2D, Charcot-Marie-Tooth neuropathy, neuronal type, D, Diadenosine tetraphosphate synthetase, DSMAV, glycine tRNA ligase, Glycine--tRNA ligase, glycyl-tRNA synthetase, GlyRSEC 6.1.1.14, HMN5, SMAD1CMT2D
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit