Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GARS Mouse anti-Human, Clone: 1641CT837.23.96, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524747

Catalog No. PIMA524747

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. The encoded enzyme is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.
Specifications

Specifications

GARS
Monoclonal
1.03 mg/mL
PBS with 0.09% sodium azide
P41250
Glycine--tRNA ligase, 6.1.1.14, Diadenosine tetraphosphate synthetase, AP-4-A synthetase, Glycyl-tRNA synthetase, GlyRS, GARS
Mouse
IgG1, kappa
200 μL
-20°C
Primary
2617
Western Blot
1641CT837.23.96
Unconjugated
GARS
Liquid
GARS
Recombit protein between 15-305 amino acids from human GARS
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit