Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GART Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | GART |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157774
|
Novus Biologicals
NBP157774 |
100 μL |
Each for $436.00
|
|
NBP15777420
|
Novus Biologicals
NBP15777420UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
GART Polyclonal specifically detects GART in Human samples. It is validated for Western Blot.Specifications
GART | |
Polyclonal | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
P22102 | |
2618 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
AIRS, EC 6.3.3.1, EC 6.3.4.13, GARS, GARTF, MGC47764, PGFTEC 2.1.2.210PAIS, phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamidesynthetase, phosphoribosylaminoimidazole synthetase, trifunctional purine biosynthetic protein adenosine-3 | |
GART | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title