Learn More
Thermo Scientific™ Gas Guards for CO2 and LN2 Tanks
Minimize the need to frequently change out cylinders and reduce the risk of running out of gas. These gas guards are factory installed and built in to specified CO2 incubators.
$538.50
Products 1
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity & Availability  
13-998-121
View Documents
Thermo Scientific™
190640
Gas Guard, N2 3130/3131, 3140/3141 incubators Each for $538.50
Add to cart
Description
Description
Minimize the need to frequently change out cylinders and reduce the risk of running out of gas. Thermo Scientific™ Gas Guards for CO2 and LN2 Tanks are factory installed and built in to specified CO2 incubators; specify when ordering the incubator. Fitting unobtrusively inside the incubator casing, these systems interface directly with unit power supply and control systems.
  • Factory-installed systems monitor CO2 or N2 pressure in source tanks
  • Automatically switch from one cylinder to another when supply in first tank is exhausted (pressure falls below 15psi)
  • Audible and visual alarms on incubator control panel alert to tank switch-overs and to when both tanks are low