Learn More
Gas1 Rat anti-Human, Alexa Fluor 405, Clone: 11C47, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP222032AF405
Catalog No. NB222032405
Add to cart
Description
Description
Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Gas1
11C47
Western Blot, ELISA
Purified
GAS1
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human Gas1. Rat anti human Gas1 NP_002039.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1
Rat
IgG2
0.1 ml
Primary
2619