Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gas1 Rat anti-Human, Alexa Fluor™ 594, Clone: 11C47, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222032AF594

Catalog No. NP222032A94


Add to cart

Description

Description

Gas1 Monoclonal antibody specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Gas1
11C47
Western Blot, ELISA
Conjugated, Purified
GAS1
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human Gas1. Rat anti human Gas1 NP_002039.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1
Rat
IgG2
0.1 mL
Primary
2619.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit