Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gas1 Rat anti-Human, Clone: 11C47, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP222032
Description
Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA.Specifications
Gas1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:1000, ELISA 1:1000-1:2000 | |
NP_002039 | |
GAS1 | |
Human recombinant Gas1 | |
0.1 mg | |
Primary | |
Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2 |
Western Blot, ELISA | |
11C47 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1 | |
Rat | |
Protein G purified | |
RUO | |
2619 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only