Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS2L2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548197

Catalog No. PA548197

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Seems to be involved in the cross-linking of microtubules and microfilaments.
Specifications

Specifications

GAS2L2
Polyclonal
PBS with 0.09% sodium azide
Q8NHY3
GAS2-like protein 2, GAS2-related protein on chromosome 17, Growth arrest-specific protein 2-like 2, GAS2L2, GAR17
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
246176
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GAS2L2
Liquid
GAS2L2
KLH conjugated synthetic peptide between 763-791 amino acids from the C-terminal region of human GAS2L2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit