Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gas6 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0303-5Y23, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212326AF532

Catalog No. NP212326A32


Add to cart

Description

Description

Gas6 Monoclonal antibody specifically detects Gas6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Gas6
MM0303-5Y23
Western Blot
Conjugated, Purified
GAS6
Human GAS 6
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6
Mouse
IgG1
0.1 mL
Primary
2621.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit