Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS7 Mouse anti-Human, Rat, Canine, PE-Cyanine7, Clone: OTI2F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271376PECY7

Catalog No. N271376PEC7


Add to cart

Description

Description

GAS7 Monoclonal antibody specifically detects GAS7 in Human, Rat, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

GAS7
OTI2F6
Flow Cytometry
Affinity Purified, Conjugated
GAS7
Full length recombinant protein of human GAS7 (NP_003635) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication
Primary
8522.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
GAS-7, growth arrest-specific 7, KIAA0394growth arrest-specific protein 7, MGC1348, MLL/GAS7, MLL/GAS7 fusion protein
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit