Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS7 Mouse anti-Human, Clone: OTI8H10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525285

Catalog No. PIMA525285


Add to cart

Description

Description

Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predomitly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described.
Specifications

Specifications

GAS7
Monoclonal
0.7 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O60861
MLL/GAS7, MLL/GAS7 fusion protein, growth arrest-specific protein 7
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8522
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI8H10
Unconjugated
GAS7
Liquid
GAS7
Full length human recombit protein of GAS7 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit