Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS8 Antibody (OTI10G2) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270795

Catalog No. NBP270795


Add to Cart

Description

Description

GAS8 Monoclonal specifically detects GAS8 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

GAS8
Monoclonal
LYOPH
Western Blot 1:2000
GAS-11, GAS11MGC138326, GAS-8, growth arrest specific 11, growth arrest-specific 11, growth arrest-specific 8, Growth arrest-specific protein 11, growth arrest-specific protein 8
Mouse
56.2 kDa
100 μg
2622
Human, Mouse, Rat
Lyophilized
Western Blot
OTI10G2
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
GAS8
Full length human recombinant protein of human GAS8 (NP_001472) produced in E.coli.
Affinity Purified
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit