Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GASP-1/WFIKKN2 Mouse anti-Human, Alexa Fluor™ 594, Clone: 17V26, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221976AF594

Catalog No. NP221976A94


Add to cart

Description

Description

GASP-1/WFIKKN2 Monoclonal antibody specifically detects GASP-1/WFIKKN2 in Human samples. It is validated for ELISA.
Specifications

Specifications

GASP-1/WFIKKN2
17V26
ELISA
Conjugated, Purified
WFIKKN2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant GASP-1. Mouse anti human WFIKKN2 Q8TEU8.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
GASP1, GASP-1, Growth and differentiation factor-associated serum protein 1, hGASP-1, WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing-relatedprotein, WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2, WAP, FS, Ig, two KU and NTR module related protein, WAP, kazal, immunoglobulin, kunitz and NTR domain-containing protein 2, WFIKKN-related protein, WFIKKNRPmultivalent protease inhibitor protein
Mouse
IgG2
0.1 mL
Primary
124857.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit