Learn More
GASP-1/WFIKKN2 Mouse anti-Human, Alexa Fluor™ 750, Clone: 17V26, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221976AF750
Catalog No. NP221976A70
Add to cart
Description
Description
GASP-1/WFIKKN2 Monoclonal antibody specifically detects GASP-1/WFIKKN2 in Human samples. It is validated for ELISA.
Specifications
Specifications
GASP-1/WFIKKN2
17V26
ELISA
GASP1, GASP-1, Growth and differentiation factor-associated serum protein 1, hGASP-1, WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing-relatedprotein, WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2, WAP, FS, Ig, two KU and NTR module related protein, WAP, kazal, immunoglobulin, kunitz and NTR domain-containing protein 2, WFIKKN-related protein, WFIKKNRPmultivalent protease inhibitor protein
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
WFIKKN2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant GASP-1. Mouse anti human WFIKKN2 Q8TEU8.
0.1 mL
Primary
124857