Learn More
Gast™ Compact Oil Free Compressor
For industrial and laboratory compressed air applications
Manufacturer:  Gast™ 1770001
Catalog No. 01-093-300
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The system utilizes an oil-less rocking piston compressor that produces clean, oil-free pressurized air flow when connected to an industrial or laboratory device. The tank ensures that a constant supply of air is available to the device.
  • 5µm filter with autodrain
  • Pressure regulator
  • Automatic drain system with evaporation tray
  • Hour run meter
  • Casters for ease of movement
  • Receiver with internal coating and 5-year warranty
  • 100% oil free
  • 100% duty cycle
  • Super quiet - only 50dBA
  • Pressure up to 7 bar
  • Long-life, 2-year warranty
  • 8000 hours cup life at 7 bar
  • Easy to use
  • Plug & play
The operation manual provides installation, operation and preventative maintenance guidelines that should be followed to ensure correct/reliable performance of this system.
Specifications
Specifications
6.4 A
60 Hz
120 V
120 V 60 Hz
7 bar