Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gast™ Oil-Less Piston-Type Pressure/Vacuum Pumps
For air sampling, pressure/vacuum filtration, degassing solutions
Manufacturer: Gast™ 1HAB25BM100X
Includes: Pump, vacuum and pressure regulators, moisture trap, intake filter, handle, rubber suction-cup feet and line cord and plug with ON/OFF switch
Description
Use with therapeutic equipment and flame photometers.
Pressure/vacuum pumps with 1/6hp motors
- Produce maximum gauge pressure of 100psig (689kPag) or vacuum of 25 in.Hg (84kPag)
- Separate vacuum and pressure gauges, graduated from 0 to 30 in.Hg (0 to 102kPag) and 0 to 100psig (0 to 689kPag), respectively
- Free-air flow: 1.3 cu. ft. (37L)/min.
Order Info
Dimensions: 15L × 8W × 10 3/4 in.H (38 × 20.3 × 27.3cm). For 115V 60Hz.
Specifications
0.25 hp | |
Pump, vacuum and pressure regulators, moisture trap, intake filter, handle, Rubber suction-cup feet and line cord and plug with on/off switch | |
37 L/min. | |
60 Hz | |
27.3 cm | |
38 cm | |
100 psi |
3.9 A | |
115 V 60 Hz | |
Therapeutic Equipment and Flame Photometers | |
10.75 in. | |
15 in. | |
8 in. | |
115 V |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title