Learn More
CPC Scientific H-Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-Arg-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GIPS002B
Encompass
GIP fragment that presents potent insulinotropic activity in the isolated perfused rat pancreas, but greatly inhibits somatostatinotropic activity in the isolated perfused rat stomach. The site responsible for insulinotropic activity lies between 19 and 30 of GIP. ONE-LETTER SEQUENCE: YAEGTFISDYSIAMDKIRQQDFVNWLLAQK-NH2MOLECULAR FORMULA: C162H245N41O47S1MOLECULAR WEIGHT:3551.04STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [134846-93-8], SYNONYMS: Gastric Inhibitory Polypeptide (1-30) amide (porcine), Glucose-Dependent Insulinotropic Polypeptide (1-30) amide (porcine), GIP (1-30) amide (porcine)RESEARCH AREA: DiabetesREFERENCES: W.J.Rossowski et al., Reg. Pep., 39, 9 (1992)
Catalog No. 50-210-9356
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.