Learn More
Alfa Aesar™ Gastrin I, Rat
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66510EXD
 View more versions of this product
Catalog No. AAJ66510EXD
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
2126.28
Rat
Solid
RUO
Gastrin I, rat
HPLC
0.5mg
Glp-Arg-Pro-Pro-Met-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2
-30° to -10°C