Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gastrin Monoclonal Antibody (C25), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $437.00
Specifications
Antigen | Gastrin |
---|---|
Clone | C25 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551079
|
Invitrogen™
MA551079 |
200 μL |
Each for $437.00
|
|
|||||
FEMA551080
|
Invitrogen™
MA551080 |
20 μL |
Each for $195.80
|
|
|||||
Description
This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.Specifications
Gastrin | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Big gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrin | |
Gast | |
IgG2a κ | |
Protein A/G | |
Gast |
C25 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P01350, P48757 | |
14459, 2520 | |
Recombinant Human GT protein, Met1-Asn101 (Accession #P01350). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title