Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin Mouse anti-Human, Clone: GAST/2632, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Gastrin
Clone GAST/2632
Concentration 0.2 mg/ml
Dilution ELISA 1-5 ug/ml
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123101
View Documents
Novus Biologicals
NBP30781410UG
100 μg
Each for $352.89
Add to cart
 
NB123102
View Documents
Novus Biologicals
NBP30781420UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Gastrin Monoclonal antibody specifically detects Gastrin in Human samples. It is validated for ELISA
Specifications

Specifications

Gastrin
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
2520
IgG1 Kappa
Primary
Store at 4°C.
GAST/2632
ELISA 1-5 ug/ml
Unconjugated
Mouse
Human
GAS, gastrin
Recombinant full-length human Gastrin protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit