Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GAT3
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094025
View Documents
Novus Biologicals
NBP303494100UL
100 μg
Each for $320.71
Add to cart
 
NB094026
View Documents
Novus Biologicals
NBP30349420UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

GAT3 Polyclonal antibody specifically detects GAT3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

GAT3
Unconjugated
Rabbit
Neuroscience
PBS with 50% glycerol, pH7.3.
6538
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse, Rat
GABA Transporter 3, GABT3, GAT-3, GAT3sodium- and chloride-dependent GABA transporter 3, GAT4, solute carrier family 6 (neurotransmitter transporter, GABA), member 11, Solute carrier family 6 member 11
Recombinant fusion protein containing a sequence corresponding to amino acids 1-60 of human GAT3 (NP_055044.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit