Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-1 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™

Rabbit Polyclonal Antibody

Manufacturer:  Active MotifSupplier Diversity Partner 61535

Encompass

 View more versions of this product

Catalog No. 50-199-3478


Add to cart

Description

Description

Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.
Specifications

Specifications

GATA-1
ChIP assay, ChIP-sequencing
Unconjugated
Gata1
Liquid
Rabbit
IgG
100 μg
-20°C, Avoid Freeze/Thaw Cycles
Primary
2623
Gata1
Lot-Specific
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
P15976
ERYF1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT
This antibody was raised against a peptide within the N-terminal region of human GATA-1.
Protein A
RUO
Antibody
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit