Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-3 Mouse anti-Human, Clone: GATA3/2442, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30766110UG
Description
GATA-3 Monoclonal specifically detects GATA-3 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
GATA-3 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary) | |
100 μg | |
Cancer, Innate Immunity | |
2625 | |
Store at 4°C. | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
GATA3/2442 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title