Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-3 Mouse anti-Human, Clone: GATA3/2442, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30766120UG

 View more versions of this product

Catalog No. NB122796


Add to cart

Description

Description

GATA-3 Monoclonal antibody specifically detects GATA-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

GATA-3
GATA3/2442
Unconjugated
10 mM PBS with 0.05% BSA
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3
Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG2b Kappa
20 μg
Cancer, Innate Immunity
Primary
2625
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit