Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-3 Mouse anti-Human, Clone: GATA3/2444, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30766220UG
Description
GATA-3 Monoclonal specifically detects GATA-3 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
GATA-3 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary) | |
20 μg | |
Cancer, Innate Immunity | |
2625 | |
Store at 4°C. | |
IgG2b κ |
Immunohistochemistry (Paraffin), Peptide Array | |
GATA3/2444 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction