Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-3 Mouse anti-Human, Clone: GATA3/2446, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen GATA-3
Clone GATA3/2446
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122882
View Documents
Novus Biologicals
NBP30770420UG
20 μg
Each for $138.37
Add to cart
 
NB122881
View Documents
Novus Biologicals
NBP30770410UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

GATA-3 Monoclonal antibody specifically detects GATA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications

Specifications

GATA-3
0.2 mg/ml
Western Blot, Flow Cytometry, ELISA
Purified
RUO
Human
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3
Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
GATA3/2446
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA
Unconjugated
Mouse
Cancer, Innate Immunity
10 mM PBS with 0.05% BSA
2625
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit