Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gata-3 Rat anti-Human, Mouse, Porcine, Rhesus Monkey, Clone: TWAJ, eBioscience™ Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 14996695

Encompass

 View more versions of this product

Catalog No. 14-996-695


Add to cart

Description

Description

The MAb TWAJ recognizes mouse and human Gata-3, a member of the Gata family of transcription factors. Gata-3 is a T cell-specific transcription factor important for thymic development and Th2 differentiation. Expression during embryonic development is found in the central nervous system, skin, mammary glands and kidney. During development, the expression of Gata-3 is essential as homozygous knock-out of Gata-3 is embryonic lethal. The Gata-3 is also essential for T cell commitment and survival. In the thymus, expression is found mainly on the CD4 single positive cells. During Th2 differentiation, Gata-3 binds to the IL-4 promoter as well as represses the expression of T-bet, thus inhibiting Th1 differentiation. Alternative splice variants have been reported especially in the MCF7 cell line. The TWAJ Human/Mouse Gata-3 antibody will recognize both forms (50 and 45 kDa) of the protein. Staining with the TWAJ Human/Mouse Gata-3 antibody requires the use of the Foxp3/Transcription Factor Staining Buffer Set.(cat 00-5523) Crossreactivity in rhesus monkeys has been published.

Gata-3 Monoclonal antibody specifically detects Gata-3 in Human, Mouse, Porcine, Rhesus Monkey samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blot
Specifications

Specifications

Gata-3
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P23771, P23772
Rat
10 mg
Primary
Human, Mouse, Porcine, Rhesus Monkey
Antibody
IgG2b κ
Immunocytochemistry, Immunofluorescence, Western Blot
TWAJ
Unconjugated
GATA3
EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3
Affinity chromatography
RUO
14462, 2625, 713840, 733631
4° C
Liquid
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit