Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-4 Rat, Biotin, Clone: Evan, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143112B

Catalog No. NBP143112B


Add to cart

Description

Description

GATA-4 Monoclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

GATA-4
Evan
Western Blot
Purified
GATA4
The immunogen for this antibody was Gata4.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
Western Blot
Biotin
PBS with 0.05% Sodium Azide
GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
Rat
IgG2a Kappa
0.1 ml
Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary
2626
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit