Learn More
GATA-4 Rat, DyLight 488, Clone: Evan, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143112G
Catalog No. NBP143112G
Add to cart
Description
Description
GATA-4 Monoclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Western Blotting.
Specifications
Specifications
GATA-4
Evan
Western Blot
Purified
GATA4
The immunogen for this antibody was Gata4.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
Rat
IgG2a Kappa
0.1 ml
Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary
2626