Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-6 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI2B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270799AF405

Catalog No. N270799A405


Add to cart

Description

Description

GATA-6 Monoclonal antibody specifically detects GATA-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

GATA-6
OTI2B3
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
GATA6
Recombinant protein fragment corresponding to amino acids 300-449 of human GATA-6 (NP_005248) produced in E.coli.
Immunogen affinity purified
Phospho Specific, Stem Cell Markers
Primary
2627.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
GATA binding protein 6, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit