Learn More
GATA3 Mouse anti-Human,Mouse, BB700, Clone: L50-823, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566643
Catalog No. BDB566643
Add to cart
Description
Description
GATA3 Monoclonal antibody specifically detects GATA3 in Human,Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
GATA3
L50-823
BB700
Mouse
25 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Human, Mouse