Learn More
GATA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 223431AP
Catalog No. 50-173-3841
Add to cart
Description
Description
Trans-acting T-cell specific transcription factor GATA-3 is a member of GATA family of transcription factors that regulates development of multiple tissues. It is an important transcription factor in regulating human Th2 cell differentiation in vivo.
Specifications
Specifications
GATA3
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23771, P23772
GATA binding factor 3, GATA binding protein 3, GATA3, HDR
Rabbit
IgG
150 μL
-20°C
Primary
14462, 2625
Western Blot
Unconjugated
GATA3
Liquid
Gata3
GATA3 Fusion Protein Ag17833
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse