Learn More
GATA5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA1103X
Catalog No. PIPA1103X
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-103X has been successfully used in Western blot applications with human samples.
GATA binding factor 5 is a transcription regulator involved in embryonic development. In addition, the protein encoded by this gene contains two GATA-type zinc fingers and is known to bind to hepatocyte nuclear factor-1alpha (HNF-1alpha). This interaction is essential for cooperative activation of the intestinal lactase-phlorizin hydrolase promoter. GATA-5 may be involved in the establishment of cardiac smooth muscle cell diversity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GATA5
Polyclonal
Unconjugated
GATA5
GATA5, GATA binding factor 5
Rabbit
Antigen affinity chromatography
RUO
140628
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Q9BWX5
GATA5
Pooled peptides RPLVRPQKRLSSSRRA and EDDSLAPGHLEFKFEPED
20 μL
Primary
Human
Antibody
IgG
Recommendations
Recommendations