Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA6 Mouse anti-Human, Clone: OTI3E2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524897

Catalog No. PIMA524897


Add to cart

Description

Description

This gene encodes a member of the GATA family of zinc-finger transcription factors that are named for the consensus nucleotide sequence they bind in the promoter regions of target genes. GATA6 is thought to be important for regulating terminal differentiation and/or proliferation and plays a critical role in endoderm development.
Specifications

Specifications

GATA6
Monoclonal
0.2 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q92908
ASD9, AVSD5, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2627
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3E2
Unconjugated
GATA6
Liquid
GATA6
Human recombit protein fragment corresponding to amino acids 300-449 of GATA6 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit