Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GATAD2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553536
Description
Immunogen sequence: NLEVPHELPT KQDGSGVKGY EEKLNGNLRP HGDNRTAGRP GKENINDEPV DMSARRSEPE RGRLTPSPDI IVLSDNEASS PRSSSRMEER LKAANLEMFK GKGIEERQQL IKQLRDELRL EEARLV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene encodes a zinc finger protein transcriptional repressor. The encoded protein is part of the methyl-CpG-binding protein-1 complex, which represses gene expression by deacetylating methylated nucleosomes. Mutations in this gene are linked to intellectual disability and dysmorphic features associated with cognitive disability.
Specifications
GATAD2B | |
Polyclonal | |
Unconjugated | |
GATAD2B | |
AL118180; C430014D17Rik; GATA zinc finger domain containing 2B; GATA zinc finger domain-containing protein 2B; GATAD2B; KIAA1150; mKIAA1150; MRD18; p66/p68; P66beta; p68; RGD1308533; transcription repressor p66 beta component of the MeCP1 complex; transcription repressor p66 component of the MeCP1 complex; transcriptional repressor p66-beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
229542, 310614, 57459 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8VHR5, Q8WXI9 | |
GATAD2B | |
Recombinant protein corresponding to Human GATAD2B. Recombinant protein control fragment (Product #RP-89356). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction