Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATD3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30372720UL
Description
GATD3A Polyclonal specifically detects GATD3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GATD3A | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
8209 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
chromosome 21 open reading frame 33, D21S2048E, ES1, GT335, HES1, HES1KNP-Ia, human HES1 protein, homolog to E.coli and zebrafish ES1 protein, 10Protein GT335, Keio novel protein I, KNPH, KNPI, KNP-I, KNPImitochondrial, Protein KNP-I | |
Recombinant fusion protein containing a sequence corresponding to amino acids 42-268 of human GATD3A (NP_004640.4). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction