Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATE-16/GABARAPL2 Rabbit anti-Human, Janelia Fluor 549, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP300320JF549

Catalog No. NB065548


Add to cart

Description

Description

GATE-16/GABARAPL2 Polyclonal antibody specifically detects GATE-16/GABARAPL2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

GATE-16/GABARAPL2
Janelia Fluor 549
50mM Sodium Borate
ATG8, ATG8C, FLC3A, gamma-aminobutyric acid receptor-associated protein-like 2, Ganglioside expression factor 2, GATE16, GATE-16, GEF2, GEF-2, GEF2GEF-2, General Protein Transport Factor P16, Golgi-Associated ATPase Enhancer Of 16 KDa, MAP1 light chain 3 related protein, MAP1 light chain 3-related protein
Produced in rabbits immunized with purified, recombinant GATE-16/GABARAPL2 (P60520; Met1-Phe117).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Autophagy, Golgi Apparatus Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
11345
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit