Learn More
Brady™ Gate Valve Lockout Pouch with Safety Padlocks & Tags
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Gate Valve Lockout Pouch with Safety Padlocks & Tags provides an easy way to lockout more valves with fewer devices. These two versatile lockouts are all that is needed to secure virtually any type and size of gate valve.
Manufacturer:  Brady™ 102692
Catalog No. 19-104-146
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
Safety
Mechanical risk lockouts
8 in.
3 in.
7 in.
1/Kit
White on red
(1) lockout bag; (1) adjustable gate valve lockout, 1-6.5 in.; (1) all purpose cable lockout; (2) kd safety padlocks, 1.5 in. shackle, red; (2) lockout tags
Nylon
3 in. x 7 in. x 8 in.