Learn More
Brady™ Gate Valve Lockout Satchel Kit w/ Brady Safety Padlocks & Tags
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Gate Valve Lockout Satchel Kit is a convienent solution that includes five sizes of gate valve lockouts in a red satchel with a shoulder strap.
Manufacturer:  Brady™ 99677
Catalog No. 19-104-197
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Mechanical risk lockouts
14 in.
15 in.
4 in.
Safety
Black on red
(1) lockout bag; (1) gate valve lockout, 1-2.5 in.; (1) gate valve lockout, 2.5-5 in.; (1) gate valve lockout, 5-6.5; in.(1) gate valve lockout, 6.5-10 in.; (1) gate valve lockout, 10-13 in.; (2) kd safety padlocks, 1.5 in. shackle, red; (2) lockout tags; (2) nylon cable ties
Nylon
15 in. x 4 in. x 14 in.