Learn More
GATM Antibody (OTI1E3), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270802G
Catalog No. NBP270802G
Add to Cart
Description
Description
GATM Monoclonal specifically detects GATM in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
GATM
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
GATM
Full length recombinant protein of human GATM (NP_001473) produced in HEK293T cells.
0.1 mL
2628.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1E3
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
AGATtransamidinase, AT, EC 2.1.4, EC 2.1.4.1, glycine amidinotransferase (L-arginine:glycine amidinotransferase), glycine amidinotransferase, mitochondrial, L-arginine:glycine amidinotransferase, Transamidinase
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG2a
Recommendations
Recommendations