Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATM Mouse anti-Human, PE-Cyanine7, Clone: OTI1E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270802PECY7

Catalog No. N270802PEC7


Add to cart

Description

Description

GATM Monoclonal antibody specifically detects GATM in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

GATM
OTI1E3
Flow Cytometry
Affinity Purified, Conjugated
GATM
Full length recombinant protein of human GATM (NP_001473) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
AGATtransamidinase, AT, EC 2.1.4, EC 2.1.4.1, glycine amidinotransferase (L-arginine:glycine amidinotransferase), glycine amidinotransferase, mitochondrial, L-arginine:glycine amidinotransferase, Transamidinase
Mouse
IgG2a
0.1 mL
Primary
2628.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit