Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORS Nasco Gatorade™ Thirst Quencher Drinks, Instant Powder Packets

Carbohydrate-electrolyte fluid-replacement drink scientifically designed for people who engage in strenuous activity

$332.12 - $420.00

Specifications

Format Powder
For Use With (Application) Provides additional energy to tired muscles and resolves hyperthermia by lowering internal body temperature
View More Specs

Products 10
Catalog Number Mfr. No. Volume (English) Volume (Metric) Flavor Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Volume (English) Volume (Metric) Flavor Quantity Price Quantity & Availability  
19-080490
View Documents
ORS Nasco
30833673
21 oz. 595 g Riptide Rush 32 Cs.
Case of 32 for $420.00
Add to cart
 
17-816-13
View Documents
ORS Nasco
30833691
21 oz. 595 g Fruit Punch 32 Cs.
Case of 32 for $397.34
Add to cart
 
17-816-14
View Documents
ORS Nasco
30803970
21 oz. 595 g Orange 32 Cs.
Case of 32 for $420.00
Add to cart
 
17-816-9
View Documents
ORS Nasco
30803957
8.5 oz. 251 mL Orange 40 Cs.
Case of 40 for $332.12
Add to cart
 
17-816-17
View Documents
ORS Nasco
30833676
51 oz. 1.4 kg Glacier Freeze 14 Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-816-10
View Documents
ORS Nasco
30803956
8.5 oz. 241 g Lemon Lime 40 Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-086826
View Documents
ORS Nasco
30833665
8.5 oz. 241 g Riptide Rush 40 Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-034363B
View Documents
ORS Nasco
30803944
21 oz. 595 g Variety Pack 32 Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-816-11
View Documents
ORS Nasco
30803808
8.5 oz. 241 g Fruit Punch 40 Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-086200
View Documents
ORS Nasco
30833672
51 oz. 1.4 kg Riptide Rush 14 Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Replaces lost fluids and electrolytes caused by heat and physical exertion
  • Provides additional energy to tired muscles, and resolves hyperthermia by lowering internal body temperature
  • Research conducted at the University of South Carolina has shown that Gatorade is quickly absorbed into the bloodstream, helping to provide rapid rehydration
  • Superior to water for enhancing physical performance during prolonged physical activity
  • Five flavors and a variety pack
  • Variety pack contains Lemon-Lime, Fruit Punch, Orange and Grape flavors

Insulated containers and coolers are also available; contact your Fisher Safety Customer Service Representative for more details.

Specifications

Specifications

Powder
Provides additional energy to tired muscles and resolves hyperthermia by lowering internal body temperature
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit