Gatorade™ Thirst Quencher Drinks, Instant Powder Packets

Carbohydrate-electrolyte fluid-replacement drink scientifically designed for people who engage in strenuous activity

Manufacturer: Nasco 308-33677

 View all options for this product

Catalog No. 17-816-16

  Check Availability
Add to Cart


    Quantity 32 Case
    Volume (English) 21 oz.
    Format Powder
    Volume (Metric) 595g
    For Use With (Application) Provides additional energy to tired muscles and resolves hyperthermia by lowering internal body temperature
    Flavor Glacier Freeze

    • Replaces lost fluids and electrolytes caused by heat and physical exertion
    • Provides additional energy to tired muscles, and resolves hyperthermia by lowering internal body temperature
    • Research conducted at the University of South Carolina has shown that Gatorade is quickly absorbed into the bloodstream, helping to provide rapid rehydration
    • Superior to water for enhancing physical performance during prolonged physical activity
    • Five flavors and a variety pack
    • Variety pack contains Lemon-Lime, Fruit Punch, Orange and Grape flavors

    Insulated containers and coolers are also available; contact your Fisher Safety Customer Service Representative for more details.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit