Gatorade™ Thirst Quencher Drinks, Instant Powder Packets

Carbohydrate-electrolyte fluid-replacement drink scientifically designed for people who engage in strenuous activity

Manufacturer: Nasco 30803968

 View more versions of this product

Catalog No. 17-816-19

  Check Availability
Add to Cart

    Specifications

    Flavor Orange
    For Use With (Application) Provides additional energy to tired muscles and resolves hyperthermia by lowering internal body temperature
    Format Powder
    Volume (English) 51 oz.
    Volume (Metric) 1.4kg
    Quantity 14 Case

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit