Learn More
Gay-Lussac's Law and Absolute Zero
Click to view available options
:
Gay-Lussacs Law and Absolute Zero
Description
Requires
- Thermometer
- Beaker
- Bulb can be submerged in boiling water, ice water, and liquid nitrogen
- Using these three widely spread temperature points versus pressure, students graph and extrapolate to Absolute Zero
Specifications
Specifications
Product Type | thermodynamic Apparatus |
For Use With (Application) | Students graph and extrapolate to absolute zero |