Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBA Mouse anti-Human, Clone: OTI1D12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526589

Catalog No. PIMA526589


Add to cart

Description

Description

GBA encodes a lysosomal membrane protein that cleaves the beta-glucosidic linkage of glycosylceramide, an intermediate in glycolipid metabolism. Mutations in this gene cause Gaucher disease, a lysosomal storage disease characterized by an accumulation of glucocerebrosides. A related pseudogene is approximately 12 kb downstream of this gene on chromosome 1. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

GBA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P04062
GBA1; GCB; GLUC; D-glucosyl-N-acylsphingosine glucohydrolase; acid beta-glucosidase; alglucerase; beta-glucocerebrosidase; glucosylceramidase; imiglucerase; lysosomal glucocerebrosidase
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2629
Immunohistochemistry (Paraffin), Western Blot
OTI1D12
Unconjugated
GBA
Liquid
GBA
Human recombit protein fragment corresponding to amino acids 40-315 of GBA produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit